Collagen I (from Rat Tail Tendons)
Supplier: ENZO LIFE SCIENCES
ALX5224400050
ENZOALX5224400050EA 120 EUR
ENZOALX5224400050
Collagen I (from Rat Tail Tendons)
Proteins and Peptides
Collagen is the main component in connective tissue and helps to provide support for tissues. It is made up of several classes, with Type 1 collagen being the most common.
Type 1 collagen has a herterotrimeric triple helical structure made up of two alpha-1(I) and one alpha-2(I) chains that form into elongated fibrils which are extremely strong. These fibrils can be found in skin, tendons, ligaments, and other connective tissues.
Type 1 collagen has been shown to be useful as a substrate that promotes cell growth and proliferation. Under acidic conditions the protein is soluble, however by raising the temperature and pH the solution forms into a solid gel that can be useful for cellular studies. It can also be dried to form a thin layer on solid surfaces such as plates, slides, or coverslips to aid in cell attachment.