Carbamazepine

Supplier: ENZO LIFE SCIENCES

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

ALX550084G005
ENZOALX550084G005EA 68.2 EUR
ENZOALX550084G005
Carbamazepine
Carbamazepine
Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
MDL Number: MFCD00005073
CAS Number: 298-46-4
EINECS: 206-062-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR