Luzindole 96

Supplier: ENZO LIFE SCIENCES

Synonyms: N-Acetyl-2-benzyl-tryptamine

ALX550184M005 ALX550184M025
ENZOALX550184M005EA 101 EUR
ENZOALX550184M005 ENZOALX550184M025
Luzindole 96
Luzindole

Competitive melatonin receptor antagonist.

Formula: C₁₉H₂₀N₂O
MW: 292.382 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672498
CAS Number: 117946-91-5

Order Now

Specification Test Results

Purity ≥96%
Appearance White to off-white crystalline solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR