N-Vanillylnonanamide [=Capsaicin (Synthetic)] ≥96% (by HPLC), white to off-white solid

Supplier: ENZO LIFE SCIENCES
Warning

Synonyms: Capsaicin (Synthetic), N-Pelargonylvanillylamide, N-Nonylvanylamide, Nonivamide, N-Pelargonic acid Vanillylamide, N-(4-Hydroxy-3-methoxybenzyl)nonanamide

ALX550239M100
ENZOALX550239M100EA 57.2 EUR
ENZOALX550239M100
N-Vanillylnonanamide [=Capsaicin (Synthetic)] ≥96% (by HPLC), white to off-white solid
N-Vanillylnonanamide [=Capsaicin (Synthetic)]

Minor constituent of hot pepper (capsicum oleoresin).


Soluble in DMSO or 100% ethanol; slightly soluble in water.

Formula: C₁₇H₂₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00017286
CAS Number: 2444-46-4

Order Now

Specification Test Results

Purity (HPLC) ≥96%
Identity Determined by NMR
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR