Fenobam ≥98% (by 1H-NMR)

Supplier: ENZO LIFE SCIENCES
Danger

Synonyms: 1-(3-Chlorophenyl)-3-(1-methyl-4-oxo-4,5-dihydro-1H-imidazol-2-yl)urea

ALX550413M005
ENZOALX550413M005EA 110 EUR
ENZOALX550413M005
Fenobam ≥98% (by 1H-NMR)
Fenobam

mGluR5 antagonist.


Potent and selective non-competitive mGluR5 antagonist that displays inverse agonist properties; blocks mGluR5 constitutive activity in vitro (IC50=87 nM).


Acts at an allosteric modulatory site shared with MPEP and binds mGluR5 with Kd values of 54 nM and 31 nM for rat and human receptors respectively. Displays anxiolytic activity following oral administration in vivo.


Soluble in DMSO or methanol.

Formula: C₁₁H₁₁ClN₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00868019
CAS Number: 57653-26-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity (¹H-NMR) ≥98%
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR