Anti-TACI Rat Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ALX804800C100
ENZOALX804800C100EA 591 EUR
ENZOALX804800C100
Anti-TACI Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For Immunohistochemistry

Recognizes mouse TACI.

Type: Primary
Antigen: TACI
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR