Matrix Metalloproteinase-1 (MMP-1) fluorometric drug discovery kit
Supplier: ENZO LIFE SCIENCES
BMLAK4050001
ENZOBMLAK4050001EA 632 EUR
ENZOBMLAK4050001
Matrix Metalloproteinase-1 (MMP-1) fluorometric drug discovery kit
Assays Enzyme Assays
A QuantiZyme™ Assay System. The MMP-1 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.
Delivery information: Kit contains MMP-1 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.