Isradipine ≥98%

Supplier: ENZO LIFE SCIENCES
Danger

BMLCA2130010
ENZOBMLCA2130010EA 119 EUR
ENZOBMLCA2130010
Isradipine ≥98%
Isradipine

Ca²⁺ channel blocker.


Potent and selective L-type calcium channel blocker. A clinically useful antihypertensive agent which has anti-atherosclerotic effects and improves endothelium-mediated NO-dependent vasodilation. Displays neuroprotective activity in a rat model of focal ischemia.


Soluble in DMSO (25 mg/ml) or 100% ethanol (15 mg/ml).

Formula: C₁₉H₂₁N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00153820
CAS Number: 75695-93-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity ≥98% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR