Human catestatin

Supplier: ENZO LIFE SCIENCES

BMLCP64200001
ENZOBMLCP64200001EA 335 EUR
ENZOBMLCP64200001
Human catestatin
Proteins and Peptides

Human Chromogranin A [352-372]). Wild type catestatin, initially described as a potent catecholamine release inhibitory peptide. Cst located in the skin is active against Gram-negative and Gram-positive bacteria as well as against fungi and yeast. Also proposed as a modulator or regulator of cardiac function and as an anti-hypertensive agent.

New Product

Order Now

SPECIFICATIONS

Protein/peptide name Catestatin
Environmentally Preferable
Species Human
Source
Purity 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR