VDM-11 ≥98% (by TLC)

Supplier: ENZO LIFE SCIENCES

Synonyms: N-Arachidonoyl-4-hydroxy-2-methylaniline, N-(4-Hydroxy-2-methylphenyl)arachidonoylamide

BMLCR1110050
ENZOBMLCR1110050EA 328 EUR
ENZOBMLCR1110050
VDM-11 ≥98% (by TLC)
VDM-11

Selective and potent inhibitor of the anandamide membrane transporter (AMT) (neuronal (C6 glioma cells): IC50=11µM; non-neuronal (rat RBL-2H3 cells): IC50=7.2µM) with negligible action at TRPV1 and weak action at CB1 receptor (CB1: Ki=5µM; CB2: Ki>5µM) and FAAH (IC50=3.7µM/ IC50>50µM (N18TG2 cell membranes).


  • Anandamide transporter inhibitor


Tan oil.


Soluble in DMSO (1.5mg/ml) or ethanol (1mg/ml).

Formula: C₂₇H₃₉NO₂
MW: 409.6 g/mol
Storage Temperature: Freezer
CAS Number: 313998-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47f5507d3313aee4fd2107be111a0563a328aeb7&vid=0900ab7e6bbe9a598a46e68bcd1f98f10c9492dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR