KT5720 ≥98% (by TLC)
Supplier: ENZO LIFE SCIENCES
BMLEI1990100
ENZOBMLEI1990100EA 186 EUR
ENZOBMLEI1990100
KT5720 ≥98% (by TLC)
KT5720
PKA inhibitor
KT5720 is a hexylester derivative of K-252a and is a selective inhibitor of PKA (Ki=0.056 µM). Cell permeable.
CAS Number: 108068-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea2d84d1dfc787149e778284183b194dbfd6d827&vid=d387b786f8a24cc74c4c67f8169bcaef58de3714&prid=web&clid=SAPDAM