SB-415286 ≥98% (by HPLC)

Supplier: ENZO LIFE SCIENCES

Synonyms: SB 415286, 3-[(3-chloro-4-hydroxyphenyl)amino]-4-(2-nitrophenyl)-1H-pyrrole-2,5-dione, 3-((3-Chloro-4-hydroxyphenyl)amino)-4-(2-nitrophenyl)-1H-pyrrole-2,5-dione

BMLEI3110005
ENZOBMLEI3110005EA 283 EUR
ENZOBMLEI3110005
SB-415286 ≥98% (by HPLC)
SB-415286

Potent and selective inhibitor of GSK-3β (IC50=78nM). Stimulates glycogen synthesis in human liver cells and mimics other actions of insulin. Protects primary neurons from death induced by PI-3 kinase pathway inhibition suggesting therapeutic potential in neurodegenerative diseases and stroke.


  • GSK inhibitor


Yellow Solid.


Soluble in DMSO, ethanol or methanol.

Formula: C₁₆H₁₀ClN₃O₅
Storage Temperature: Freezer
MDL Number: MFCD04039789
CAS Number: 264218-23-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed37859e50a544d206b2ce4aed524b060d1cdee&vid=67f0a2839edda0944e77b6554b20e74c0340da9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR