Nicotinamide

Supplier: ENZO LIFE SCIENCES

Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3

BMLKI2830500
ENZOBMLKI2830500EA 50.6 EUR
ENZOBMLKI2830500
Nicotinamide
Nicotinamide

Nicotinamide, the water-soluble amide of nicotinic acid, is a component of the two most important coenzymes-nicotinamide adenine dinucleotide and nicotinamide adenine dinucleotide phosphate. Nicotinamide is involved in numerous oxidation-reduction reactions in mammalian biological systems acting as an antioxidant.


Soluble in water, 100% ethanol or glycerol. Slightly soluble in chloroform, almost insoluble in ether or acetone.

Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 334
Flash Pt: 182
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
Merck Index: 14,06523
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28defecca22e1ab24aa0e7cbff11f6c6258a1a42&vid=d30f56d9f85db795ac8eeaad53d4fc6aa9ec2a8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation 50 mM nicotinamide in 50 mM tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl₂.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR