Src substrate II

Supplier: ENZO LIFE SCIENCES

BMLP2590001
ENZOBMLP2590001EA 92.4 EUR
ENZOBMLP2590001
Src substrate II
Proteins and Peptides

This peptide, YIYGSFK, was derived through a random combinatorial peptide library method and is a specific and efficient substrate for the scr family of kinases. The Km of YIYGSFK for p60c-src is 55 µM.

Order Now

SPECIFICATIONS

Protein/peptide name Src Substrate II
Environmentally Preferable
Species
Source
Purity >95% (HPLC)
Formulation Lyophilised

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR