Castanospermine 98.5-102.0%

Supplier: ENZO LIFE SCIENCES

Synonyms: (1S,6S,7R,8R,8aR)-Octahydroindolizine-1,6,7,8-tetraol

BMLS1070100 BMLS1070500
ENZOBMLS1070100EA 137 EUR
ENZOBMLS1070100 ENZOBMLS1070500
Castanospermine 98.5-102.0%
Castanospermine

Plant alkaloid. Potent inhibitor of α- and β-glucosidases. Exhibits antiviral properties. Inhibits thyroglobulin secretion. Antitumor activity. Inhibits protein glycosylation. Prevents angiogenesis.

Formula: C₈H₁₅NO₄
MDL Number: MFCD00017555
CAS Number: 79831-76-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f47db84f17b5ae5606e869d581469fff5b20c2b1&vid=60ede12dc16d11a57facc6ddcf803356bf96f5c2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR