Cyanine 3-UTP (enhanced)
Supplier: ENZO LIFE SCIENCES
ENZ42505
ENZOENZ42505EA 467 EUR
ENZOENZ42505
Cyanine 3-UTP (enhanced)
Nucleic Acid Reagents Nucleotides
Cyanine 3-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP .