Fluorescein-12-UTP
Supplier: ENZO LIFE SCIENCES
ENZ42834
ENZOENZ42834EA 478 EUR
ENZOENZ42834
Fluorescein-12-UTP
Protein Linking and Labeling Systems
Fluorescein-12-UTP is ideal for in vitro transcription reactions catalysed by T3, T7, or SP6 RNA polymerases and reverse transcriptase to produce fluorophore-labeled RNA transcripts. RNA-based probes can be used to identify complementary sequences by in situ hybridisation to fixed cells and tissues by direct or indirect fluorescence detection or by membrane hybridisation procedures.