Red detection kit (GFP-CERTIFIED®), LYSO-ID®
Supplier: ENZO LIFE SCIENCES
ENZ510050100
ENZOENZ510050100EA 165 EUR
ENZOENZ510050100
Red detection kit (GFP-CERTIFIED®), LYSO-ID®
Assays Cellular Assays
LYSO-ID® Red Detection Kit (GFP-CERTIFIED®) is an acidic organelle-selective dye for live cell staining of lysosomes. It is an ideal choice for fluorescence co-localisation imaging with GFP-tagged proteins and providing spectrally pure red signal.
  • No photo conversion to a green emitting dye
  • No complicating metachromatic dual emission artifacts, facilitating analysis with GFP, FITC and other fluorescent probes
  • Highly resistant to photobleaching and concentration quenching
  • Ensures strong and consistent fluorescence signal, even after extended viewing periods
LYSO-ID® Red Detection Kit is for use with GFP-expressing cell lines, as well as cells expressing blue, cyan or yellow fluorescent proteins.
Delivery information: Kit contains LYSO-ID® Red Detection Reagent, Hoechst 33342 nuclear stain, chloroquine control and 10X assay buffer.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable