Anti-TAU Mouse Monoclonal Antibody [clone: Tau 12]

Supplier: ENZO LIFE SCIENCES
ENZABS2160500
ENZOENZABS2160500EA 2610 EUR
ENZOENZABS2160500
Anti-TAU Mouse Monoclonal Antibody [clone: Tau 12]
Antibodies
Recommended Applications: ELISA, IHC, WB

Species reactivity: Human

Type: Primary
Antigen: TAU
Clonality: Monoclonal
Clone: Tau 12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR