TG100115 ≥98% (by HPLC)

Supplier: ENZO LIFE SCIENCES

ENZCHM1390005 ENZCHM1390010
ENZOENZCHM1390005EA 304 EUR
ENZOENZCHM1390005 ENZOENZCHM1390010
TG100115 ≥98% (by HPLC)
TG100115

Inhibitor of PI3Kγ, PI3Kδ, FRAP, Akt and VEGF.

Formula: C₁₈H₁₄N₆O₂
MDL Number: MFCD16038873
CAS Number: 677297-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=281b506299862e53db7fe5a6005230d4220969a4&vid=6307eaab5db81413045b5b2db330f827e1bba791&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR