Geno-ElectroPhore™ Mini Horizontal Gel Electrophoresis System
Supplier: G-Biosciences
BT105-C BT105-A BT105-F
GENOBT105-CEA 144 EUR
GENOBT105-C GENOBT105-A GENOBT105-F
Geno-ElectroPhore™ Mini Horizontal Gel Electrophoresis System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
For a variety of agarose electrophoresis applications, the ultimate in simplicity and convenience in casting agarose gels.
  • Eliminates gel leaking issues, no taping required
  • Cast two gel sizes: two 5×8,3 cm and one 10,5×8,3 cm gels
  • Designed for easy well identification
  • Two combs provided for small and large wells
  • Supplied with optional power supply
The casting trays are provided with background black strips for high well visibility. The combs provide either 13 and six wells (0,6×0,1 cm well) or 17 and eight wells (0,4×0,1 cm well) for the large andMore Product Information