4-Nitrophenyl acetate, pale yellow powder

Supplier: MP Biomedicals

0210088825 0210088805
ICNA0210088825EA 150 EUR
ICNA0210088825 ICNA0210088805
4-Nitrophenyl acetate, pale yellow powder
4-Nitrophenyl acetate

p-Nitrophenyl Acetate used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.


p-Nitrophenyl Acetate used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.


Soluble in ethanol (100 mg/ml), with heat as needed, yielding a clear, light yellow to yellow solution & methanol(10 mg/ml).

Formula: C₈H₇NO₄
Melting Pt: 75…79 °C (lit.)
MDL Number: MFCD00007326
CAS Number: 830-03-5
EINECS: 212-593-5

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
UV/Vis: l max (methanol) 265 - 269 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR