ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥98%

Supplier: MP Biomedicals

Synonyms: NADPH tetrasodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1(4H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl phosphate, Nicotinamide adenine dinucleotide phosphate tetrasodium salt, (reduced form)

0210116725 0210116750 0210116780 0210116783 0210116791
ICNA0210116725EA 126 EUR
ICNA0210116725 ICNA0210116750 ICNA0210116780 ICNA0210116783 ICNA0210116791
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form) ≥98%
ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)

Prepared by chemical reduction of NADP.


  • White powder


Store at −0 °C, desiccate, protect from light, store under nitrogen

Formula: C₂₁H₂₆N₇Na₄O₁₇P₃
MW: 833.35 g/mol
MDL Number: MFCD00036263
CAS Number: 2646-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR