Cholesterol, white powder NF

Supplier: MP Biomedicals

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

0210138005 02101380.1 02101380.3 02101380.5 0210138001
ICNA0210138005EA 2746 EUR
ICNA0210138005 ICNA02101380.1 ICNA02101380.3 ICNA02101380.5 ICNA0210138001
Cholesterol, white powder NF
Cholesterol

Cholesterol is the principal sterol of the higher animals, found in all body tissues. It is the main constituent of gallstones and all biological membranes. Approximately 25% of total brain lipid is cholesterol.


Cholesterol, followed by the chemical suffix -ol for an alcohol, is an organic chemical substance classified as a waxy steroid of fat.
Cholesterol is suitable for use in liposome preparations. Cholesterol is a component used for the preparation and study of artificial model membranes. One example is a study of valinomycin induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions. Positively charged cyanine dyes were used that fluorimetrically respond to the change in potential.
Major component of all biological membranes; ~25% of total brain lipid is cholesterol. It is an essential structural component of mammalian cell membranes and is required to establish proper membrane permeability and fluidity. In addition to its importance within cells, cholesterol also serves as a precursor for the biosynthesis of steroid hormones, bile acids, and vitamin D. Cholesterol is the principal sterol synthesized by animals; in vertebrates it is formed predominantly in the liver. Small quantities are synthesized in other cellular organisms (eukaryotes) such as plants and fungi. It is almost completely absent among prokaryotes (i.e., bacteria).


Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Formula: C₂₇H₄₆O
Boiling Pt: 360 °C (lit.)
Melting Pt: 147…150 °C
Density: 1,067 g/mL (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now

Specification Test Results

Identity Test A Passes
Identity Test B Passes
Solubility in Alcohol Passes
Loss on Drying <0.3%
Optical rotation (c=2, dioxane) -34° to -38°
Acidity <0.3mL
Residue on Ignition <0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR