L(+)-Citrulline, white powder

Supplier: MP Biomedicals

0210139725 0210139790 0210139791 0210139705 0210139780
ICNA0210139725EA 133 EUR
ICNA0210139725 ICNA0210139790 ICNA0210139791 ICNA0210139705 ICNA0210139780
L(+)-Citrulline, white powder
L(+)-Citrulline

Citrulline (an organic compound) is an α-amino acid. It is prepared from arginine with a concomittant release of nitric oxide. In recent studies, citrulline has been found to relax blood vessels. Circulating citrulline concentration is, in humans, a biomarker of intestinal functionality.


Citrulline is an essential constituent of antigenic determinants recognized by rheumatoid arthritis -speciï¬c autoantibodies. It has also found its application in Immunohistochemistry and ELISA.


Intermediate in the production of nitric oxide from L-arginine by NO synthetase.


Soluble in water (10% w/v - clear, colorless solution).

Formula: C₆H₁₃N₃O₃
Melting Pt: 220…221 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00064397
CAS Number: 372-75-8
EINECS: 206-759-6

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (10% aq soln) 6,0 ± 0,5
Loss on Drying 1,5%
Optical Rotation 24 ° ±5 ° (c=1, 1 N HCl)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR