6-Aminocaproic acid, powder_white

Supplier: MP Biomedicals
Warning

Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

0210164591 0210164525 0210164580 0210164590
ICNA0210164591EA 616 EUR
ICNA0210164591 ICNA0210164525 ICNA0210164580 ICNA0210164590
6-Aminocaproic acid, powder_white
6-Aminocaproic acid

ε-Amino-n-caproic acid is a plasmin inhibitor. It inhibits carboxypeptidase B. It promotes rapid dissociation of plasmin by inhibiting the activation of plasminogen. As an inhibitor of plasmin it is has been utilised in the clotting buffer for fibrinogen assays.


EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carbo­xy­peptidase, plasmin, and plasminogen activator. Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.


Soluble in water (50 mg/ml); sparingly soluble in alcohol and methanol; practically insoluble in ether; insoluble in ethanol.


Caution: Research Use Only (RUO).

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 257
Melting Pt: 127
Density: 1.48 g/mL (lit.)
Flash Pt: 207
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2
Merck Index: 13,00431

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 205 - 215 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR