2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥99%, yellow powder

Supplier: MP Biomedicals
Warning

0210206625 0210206605 02102066.1 02102066.5 0210206601
ICNA0210206625EA 1803 EUR
ICNA0210206625 ICNA0210206605 ICNA02102066.1 ICNA02102066.5 ICNA0210206601
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥99%, yellow powder
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride

p-Iodonitrotetazolium Violet is used as a electron acceptor for the colorimetric assay of various dehydrogenases.


p-Iodonitrotetazolium Violet is used as a electron acceptor for the colorimetric assay of various dehydrogenases.


Soluble in 5% methanol/water(1:1)(clear yellow); slightly soluble in water, alcohol, DMSO.

Formula: C₁₉H₁₃ClIN₅O₂
Melting Pt: 227…229 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011961
CAS Number: 146-68-9
EINECS: 205-676-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR