L(+)-Rhamnose monohydrate ∼99%, powder

Supplier: MP Biomedicals

Synonyms: (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate

0210280905 0210280910 0210280925 0210280950 0210280980
ICNA0210280905EA 56.5 EUR
ICNA0210280905 ICNA0210280910 ICNA0210280925 ICNA0210280950 ICNA0210280980
L(+)-Rhamnose monohydrate ∼99%, powder
L(+)-Rhamnose monohydrate

Soluble in water (100 mg/ml - clear, colorless solution).


Soluble in water (100 mg/ml - clear, colorless solution)


Caution: Research Use Only (RUO).

Formula: C₆H₁₂O₅·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149363
CAS Number: 10030-85-0

Order Now

Specification Test Results

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 91 - 101 °C
Moisture Report Result
Optical Rotation (c=4, H₂O, NH₃) +8.2 ± 1°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR