Thiamine hydrochloride, powder

Supplier: MP Biomedicals

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

0210302825 0210302880 0210302883 0210302891
ICNA0210302825EA 31.1 EUR
ICNA0210302825 ICNA0210302880 ICNA0210302883 ICNA0210302891
Thiamine hydrochloride, powder
Thiamine hydrochloride

Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.


Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement.


Soluble in HCl (50 mg/ml - clear, colorless solution).


Caution: Research Use Only (RUO).

Formula: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Melting Pt: 252
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
Merck Index: 13,09366

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 2,7 - 3,4
OD400 (1% aq soln) ≤0.25
Loss on Drying ≤5,0%
Residue on Ignition ≤0,2%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR