Tricaprin, solid_white

Supplier: MP Biomedicals

Synonyms: Tridecanoin, Trisdecanoin, 1,2,3-Tridecanoylglycerol, Glycerol Tridecanoate, Propane-1,2,3-triyl tris(decanoate)

0210529701 0210529705
ICNA0210529701EA 74.6 EUR
ICNA0210529701 ICNA0210529705
Tricaprin, solid_white
Tricaprin

Soluble in chloroform (100 mg/ml) and ethanol.


Soluble in ethanol.


Caution: Research Use Only (RUO).

Formula: C₃₃H₆₂O₆
MW: 554.85 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036239
CAS Number: 621-71-6

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR