di-Potassium hydrogen phosphate, anhydrous ≥98%, white powder

Supplier: MP Biomedicals

0215194505 02151945.1 02151945.5 0215194501
ICNA0215194505EA 500 EUR
ICNA0215194505 ICNA02151945.1 ICNA02151945.5 ICNA0215194501
di-Potassium hydrogen phosphate, anhydrous ≥98%, white powder
di-Potassium hydrogen phosphate

Soluble in water (1g/20 ml); Insoluble to slightly soluble in ethanol; insoluble in alcohol.

Formula: K₂HPO₄
Melting Pt: 340 °C
Density: 2,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR