5-(2-Aminoethylamino)-1-naphthalenesulphonic acid, grey powder

Supplier: MP Biomedicals
Warning

Synonyms: 5-((2-Aminoethyl)amino)naphthalene-1-sulphonic acid, EDANS, acid

0215474001
ICNA0215474001EA 83.7 EUR
ICNA0215474001
5-(2-Aminoethylamino)-1-naphthalenesulphonic acid, grey powder
5-(2-Aminoethylamino)-1-naphthalenesulphonic acid

Soluble in DMSO and 1N NaOH (2,0 mg/ ml).


Soluble in DMSO and 1N NaOH (2,0 mg/ ml).


Caution: Research Use Only (RUO).

Formula: C₁₂H₁₄N₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD00066932
CAS Number: 50402-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR