Manganese(II) chloride tetrahydrate, crystalline powder

Supplier: MP Biomedicals
Warning

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

0215533480 0215533490
ICNA0215533480EA 42.3 EUR
ICNA0215533480 ICNA0215533490
Manganese(II) chloride tetrahydrate, crystalline powder
Manganese (II) chloride tetrahydrate

Manganese chloride is used as a source of manganese ion in biological research, such as in studies of manganese uptake in rat liver and of inhibition of Mycoplasma strain growth.


The alteration in gene expression of glutamate/aspartate transporter, taurine transporter, and metallothionein I in astrocytes that have been exposed to MnCl2 has been studied. Manganese chloride has been substituted for magnesium chloride to investigate alterations in activity of the Herpes simplex virus type 1 DNA polymerase. It is used in industrial applications such as dyeing and disinfecting, the purification of natural gas, and as a drying agent for linseed oil.


Manganese chloride occurs in nature as the mineral scacchite. It is used in industrial applications such as dyeing and disinfecting, the purification of natural gas, and as a drying agent for linseed oil. In biochemistry, manganese is a cofactor in restriction enzymes such as BamH I and EcoR V.


In biochemistry, manganese is a cofactor in restriction enzymes such as BamH I and EcoR V. MnCl2 has been shown to inhibit choline transport at the blood brain barrier in rats.


Soluble in water (130 mg/ml), alcohol; insoluble in ether.

Formula: MnCl₂·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR