Glycerine ≥99%, clear colourless viscous liquid, highly purified

Supplier: MP Biomedicals

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

0219520480 0219520490 0219520491
ICNA0219520480EA 37.5 EUR
ICNA0219520480 ICNA0219520490 ICNA0219520491
Glycerine ≥99%, clear colourless viscous liquid, highly purified
Glycerine

Glycerol is a simple polyol compound which is a viscous liquid that is widely used in pharmaceutical formulations. It has three hydroxyl groups that are responsible for its solubility in water and its hygroscopic nature.


It is a trihydroxy sugar alcohol that is an intermediate in carbohydrate and lipid metabolism. It is used as a solvent, emollient, pharmaceutical agent, sweetening agent and is of low toxicity.


Glycerol is used in the concentration and storage of enzymes. Also prevents back-diffusion and protein samples into the buffer. It is an osmotic agent with lubricating and moisturizing properties and has a long list of applications. It is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component.


Soluble in aqueous solution (1 mg/ml-clear, colorless solution).

Formula: HOCH₂CH(OH)CH₂OH
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Glycerin; Glycerine; 1,2,3-Propanetriol; Glyceritol; Glycyl alcohol; Grocolene; Osmoglyn; Trihydroxypropane; 1,2,3-Trihydroxypropane
Form Clear and colorless, syrupy odorless liquid
Storage Store at Room Temperature (15 - 30 °C)
Heavy Metals 5 ppm((as Pb)
pH 5,0 - 7,5 (1% aq soln)
Refractive Index n20/D 1,4746 (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR