Sodium dihydrogen phosphate, anhydrous ≥98%, white powder

Supplier: MP Biomedicals

0219550080 0219550090 0219550091 0219550092
ICNA0219550080EA 25.8 EUR
ICNA0219550080 ICNA0219550090 ICNA0219550091 ICNA0219550092
Sodium dihydrogen phosphate, anhydrous ≥98%, white powder
Sodium dihydrogen phosphate

Sodium phosphate monobasic is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography.


Useful in conjunction with Sodium Dibasic Phosphate, in the preparation of biological buffers. Used in many applications including the purification of antibodies.


Freely soluble in water; insoluble in ethanol.


Caution: Research Use Only (RUO).

Formula: NaH₂PO₄
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
Moisture Content ≤1%
pH (1% aq soln) 3 - 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR