Glycerine ≥99.5%, Ultrapure

Supplier: MP Biomedicals

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

04800687 04800689
ICNA04800687EA 97.2 EUR
ICNA04800687 ICNA04800689
Glycerine ≥99.5%, Ultrapure
Glycerine

Mercury: <10ppm Lead: <5 ppm
Iron: <5 ppm Zinc: <2 ppm
Used in the concentration and storage of enzymes. Also prevents back-diffusion and protein samples into the buffer.


pH: 5.0 -7.5 (1% aq soln)

Formula: HOCH₂CH(OH)CH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR