Acrylamide/Bis-Acrylamide, LIQUI-GEL™ solution 40% (19:1), MP Biomedicals
Supplier: MP Biomedicals
04800803
ICNA04800803EA 250 EUR
ICNA04800803
Acrylamide/Bis-Acrylamide, LIQUI-GEL™ solution 40% (19:1), MP Biomedicals
Electrophoresis Gels Agarose and Acrylamide
Liqui-Gel is a 40% (w/v) solution of ultra pure acrylamide and bis-acrylamide in deionised water. The gel is formed by the polymerisation of the acrylamide monomer with a co-monomer often N,N'-Methylene-bis-acrylamide, which acts as a cross-linking agent, in the presence of free radicals or a photochemical initiator.
  • Clear and colourless
  • Used in protein and nucleic acid electrophoresis
Order Now
SPECIFICATIONS
Environmentally Preferable