Sodium dodecyl sulphate (SDS) ≥99%, fine white powder, Ultrapure

Supplier: MP Biomedicals
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

04811030 04811032 04811033 04811034 04811036
ICNA04811030EA 509 EUR
ICNA04811030 ICNA04811032 ICNA04811033 ICNA04811034 ICNA04811036
Sodium dodecyl sulphate (SDS) ≥99%, fine white powder, Ultrapure
Sodium dodecyl sulphate (SDS)

SDS is an anionic detergent and wetting agent that is effective in both acid and alkaline solutions.
Used to solubilize and denature proteins for denaturing-PAGE. Most proteins bind SDS in a ratio of 1.4 g SDS per gram of protein. The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS. The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on their size.


  • Ultra Pure Grade
  • Suitable for Electrophoresis
  • Presentation: Fine White Powder
  • Aggregation Number: 62(Lit.)
  • Critical Micelle Concentration (CMC): 7-10 mM (Lit.)
  • Soluble in water (200 mg/mL - clear, faint yellow solution), and ethanol (0.1g/10 mL).
  • Store at Room temperature(15-30 °C)


pH: 6 - 7 (1% aq soln)

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 180…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 180-210°C
OD260 (1% aq soln) ≤0.1
OD280 (1% aq soln) ≤0.1
pH (1% aq soln) 6-7
Heavy Metals (as Pb) ≤20 ppm
Phosphate ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR