Cetrimonium bromide, powder

Supplier: MP Biomedicals
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

0520823280
ICNA0520823280EA 46.8 EUR
ICNA0520823280
Cetrimonium bromide, powder
Cetrimonium bromide

Cetyltrimethylammonium bromide (CTAB) is used for the isolation of plant high molecular weight DNA (by a rapid method) as well as plant DNA for use in PCR analysis and to precipitate nucleic acids.


  • Ready for CE IVD labeling of clinical applications
  • Research use only (RUO)


CTAB provides enhancement of concanavalin A mediated agglutination. It can be used for the determination of protein molecular weights in electrophoretic systems and for determination of critical micelle concentration of detergents and also used as a titrant for potentiometric titration of perchlorate and as a phase-transfer catalyst in reduction of arenes and heterocyclic compounds.

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Flash Pt: 244
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
Merck Index: 13,02034

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR