DNA purification kits, Geneclean® kit, MP Biomedicals
Supplier: MP Biomedicals
111001200
ICNA111001200EA 261 EUR
ICNA111001200
DNA purification kits, Geneclean® kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The GeneClean® Kit is designed to purify fragments of DNA from 200 bp to 20 kb from solutions and agarose gels.
Applications include desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearised from uncut vector, and isolation of PCR products away from genomic DNA.
The kit simplifies the process of purifying DNA into three easy steps: Bind, wash and elute. Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications.
Accessories information: Geneclean® Glassmilk™ is a specially prepared aqueous suspension of proprietary silica matrix that allows flexibility of reaction size and volume. Geneclean® Glassmilk™ binds to radioactive isotope-labeled ssDNA or dsDNA purified from agarose resulting in purified DNA suitable for further use.
Delivery information: Supplied with Geneclean® Glassmilk™, which is also available separately.