Glycerine solution 80%, sterile

Supplier: MP Biomedicals

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

113055034 113055044
ICNA113055034EA 84.9 EUR
ICNA113055034 ICNA113055044
Glycerine solution 80%, sterile
Glycerine

Sterile 80% Glycerol solution offered is less viscous and more convenient to pipette than 100% glycerol.


Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component. It has been used in purification of bacteriophage λ particles by step gradients, calcium-phosphate-mediated transfection of eukaryotic cells with plasmid DNA, DEAE-dextranmediated transfection of DNA into cells, DNA sequencing reactions, and mediation of mispriming in PCR. The use of glycerol in improved focusing of alkaline pH protein samples in immobilized pH gradient (IPG) strips has been published. Studies of the effect of glycerol on separation of DNA fragments by capillary electrophoresis have been reported.

Formula: HOCH₂CH(OH)CH₂OH
Boiling Pt: 182 °C (20 mmHg)) (lit.)
Density: 1,25 g/mL (lit.)
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals ≤2 ppm
Water Content ≤0,5%
Storage Conditions Store at Room Temperature (15-30 °C)
Assay ≥99,5% (GC), corrected for water
Concentration 0,8
Auto ignition 698 °F (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR