Genomic DNA purification kits, Fast DNA™ kit, MP Biomedicals
Supplier: MP Biomedicals
116540400
ICNA116540400EA 435 EUR
ICNA116540400
Genomic DNA purification kits, Fast DNA™ kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The FastDNA™ Kit quickly and efficiently isolates high quality genomic DNA from plants, animals, bacteria, yeast, algae, and fungi using a silica spin filter method. It can be used with the FastPrep®-24 or FastPrep® FP120 instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.
Lysing Matrix A and three different chaotropic buffers are provided for the optimal homogenisation of a wide variety of sample types. The released DNA is purified by a silica-based GeneClean® SPIN procedure, with supplied filter buckets and catch tubes to simplify liquid handling. Isolated DNA is suitable for PCR analysis and other downstream applications.
Delivery information: The kit includes Lysing Matrix A (100× 2 ml tubes), ¼" ceramic spheres (100 spheres), Binding Matrix (66 ml), concentrated SEWS-M (12 ml), DES (25 ml), CLS VF (90 ml), PPS (25 ml), CLS-TC (110 ml) and CLS-Y (110 ml).