1-Butanol

Supplier: MP Biomedicals
Danger

19400191
ICNA19400191EA 97.1 EUR
ICNA19400191
1-Butanol
1-Butanol
Formula: H₃C(CH₂)₂CH₂OH
Boiling Pt: 117,6 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0,812 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR