CHAPS lysis buffer

Supplier: Thermo Fisher Scientific

J60580.ADEA 107 EUR
J60580.AD J60580.AE
CHAPS lysis buffer
Buffers Extraction Buffers

For cell lysis and protein extraction.


Appearance: Clear liquid
MDL No.: MFCD00282866


Contains 50 mM Tris-HCl (pH 7,4), 110 mM NaCl, 5 mM EDTA and 1% CHAPS

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR