Stoppers, universal caps, FitsAll™
Supplier: Simport Scientific
T403GY T403G T403Y T403BK T403B T403L T403N T403R
SIMPT403GYEA 44.5 EUR
SIMPT403GY SIMPT403G SIMPT403Y SIMPT403BK SIMPT403B SIMPT403L SIMPT403N SIMPT403R
Stoppers, universal caps, FitsAll™
Stoppers PE/LDPE/HDPE Stoppers
Designed for easy-on, easy-off use. Cap removal is simple and quick. FitsAll™ closure is ideal for recapping most 12 and 16 mm O.D. evacuated glass blood collection tubes. Flexible FitsAll™ closures protect from aerosols of highly infectious microorganisms. They guard samples against cross-contamination and laboratory work areas against infection and spillage. Precision molded from low-density polyethylene, FitsAll™ fits firmly on the tube. Not suitable for autoclaving.
  • PE
  • Very sturdy under most rigorous procedures, including centrifugation
  • Available in 8
More Product Information