Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanine)) [clone: Hit3a]

Supplier: Tonbo Biosciences
20-0039-T025 20-0039-T100
20-0039-T025EA 35.2 EUR
20-0039-T025 20-0039-T100
Anti-CD3 Mouse Monoclonal Antibody (APC (Allophycocyanine)) [clone: Hit3a]
Antibodies
The Hit3a antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing cell proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.

The Hit3a antibody is a widely used phenotypic marker for human T cells. In addition, binding/cross-linking of Hit3a antibody to CD3e can induce cell activation. The antibody has also been demonstrated to be cross-reactive with Chimpanzee CD3. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: Hit3a
Conjugation: APC (Allophycocyanine)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR