Cholesterol hydrogen succinate, powder

Supplier: MP Biomedicals

Synonyms: Succinic Acid Monocholesterol Ester, 5-Cholesten-3β-ol 3-hemisuccinate, Cholesteryl Hydrogen Succinate, 3β-Hydroxy-5-cholestene 3-hemisuccinate, Cholesteryl Hemisuccinate, CHS

0210137980 0210137905 0210137925
ICNA0210137980EA 2365 EUR
ICNA0210137980 ICNA0210137905 ICNA0210137925
Cholesterol hydrogen succinate, powder
Cholesterol hydrogen succinate

Appearance in solution: (5% in toluene) clear, water white

Formula: C₃₁H₅₀O₄
MW: 486,74 g/mol
MDL Number: MFCD00037705
CAS Number: 1510-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR