2,5-dioxopyrrolidin-1-yldodecanoate, lyophilised powder
Supplier: MP Biomedicals
0210481705
0210481701
ICNA0210481705EA
486
EUR
ICNA0210481705
ICNA0210481701
2,5-dioxopyrrolidin-1-yldodecanoate, lyophilised powder
2,5-dioxopyrrolidin-1-yldodecanoate
Lauric acid N-hydroxysuccinimide ester may be used for the synthesis of N-acylamino acids and lipidisation of polypeptides or other applications that use N-hydroxysuccinimide ester conjugation chemistry.
- Soluble in methanol (10 mg/ml-clear, colourless solution)
Formula:
C₁₆H₂₇NO₄ |
MDL Number:
MFCD00050401 CAS Number: 14565-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...