Citrinin, yellow, crystalline powder

Supplier: MP Biomedicals

Synonyms: Citrinin, yellow, (3S,4R)-8-hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid

0215070001 0215070005 0215070010
ICNA0215070001EA 54 EUR
ICNA0215070001 ICNA0215070005 ICNA0215070010
Citrinin, yellow, crystalline powder
Citrinin

Soluble in alcohol, dioxane, dilute alkali, dichloromethane (10 mg/mL - clear, yellow solution), practically insoluble in water.

Formula: C₁₃H₁₄O₅
MW: 250,25 g/mol
MDL Number: MFCD00006912
CAS Number: 518-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR