cis-Zeatin, powder
Supplier: MP Biomedicals
0215227105
0215227110
0215227150
ICNA0215227105EA
644
EUR
ICNA0215227105
ICNA0215227110
ICNA0215227150
cis-Zeatin, powder
cis-Zeatin
Soluble in glacial acetic acid (2 mg + 0,2 ml, clear faint yellow solution).
May contain up to 10% trans-isomer
Formula:
C₁₀H₁₃N₅O |
CAS Number:
32771-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...